This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf04383427_m1
See other SLCO1A2 GE Assays ›
Gene Symbol:
SLCO1A2
Gene Name:
solute carrier organic anion transporter family member 1A2
Gene Aliases:
-
Chromosome Location:
Chr.11: 21976025 - 22147377 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf04383427_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLCO1A2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005570311.2 XP_005570368.1 16 - 17 2241 69
XM_005570312.2 XP_005570369.1 14 - 15 1893 69
XM_005570313.2 XP_005570370.1 16 - 17 2171 69
XM_005570314.2 XP_005570371.1 15 - 16 2127 69
XM_005570315.2 XP_005570372.1 16 - 17 2255 69
XM_005570316.2 XP_005570373.1 12 - 13 4052 69
XM_005570317.2 XP_005570374.1 13 - 14 1786 69
XM_005570318.2 XP_005570375.1 14 - 15 1907 69
XM_005570319.2 XP_005570376.1 14 - 15 2119 69
XM_005570320.2 XP_005570377.1 15 - 16 2122 69
XM_005570321.2 XP_005570378.1 13 - 14 2005 69
XM_005570322.2 XP_005570379.1 12 - 13 1982 69
XM_005570323.2 XP_005570380.1 13 - 14 1774 69
XM_005570324.2 XP_005570381.1 12 - 13 1919 69
XM_005570325.2 XP_005570382.1 12 - 13 1752 69
XM_005570327.2 XP_005570384.1 11 - 12 2351 69
XM_015430466.1 XP_015285952.1 14 - 15 1955 69
XM_015430467.1 XP_015285953.1 14 - 15 1948 69
XM_015430468.1 XP_015285954.1 13 - 14 1834 69
XM_015430469.1 XP_015285955.1 12 - 13 2221 69
XM_015430470.1 XP_015285956.1 12 - 13 2141 69
XM_015430471.1 XP_015285957.1 12 - 13 1644 69
XM_015430472.1 XP_015285958.1 14 - 15 1980 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

transporter

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0