Assay Details
| Gene Symbol: KRT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005570931.2 | XP_005570988.1 | 8 - 9 | 1588 | 77 | |
| XM_015430960.1 | XP_015286446.1 | 9 - 10 | 1480 | 77 | ||
More Information
Set Membership: |
|

KRT3
Genomic Map