This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf04385030_m1
See other DTX2 GE Assays ›
Gene Symbol:
DTX2
Gene Name:
deltex E3 ubiquitin ligase 2
Gene Aliases:
-
Chromosome Location:
Chr.3: 46936371 - 46982470 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf04385030_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DTX2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001319408.1 NP_001306337.1 2 - 3 453 53
XM_005549324.1 XP_005549381.1 2 - 3 534 53
XM_005549325.2 XP_005549382.1 3 - 4 680 53
XM_015447202.1 XP_015302688.1 3 - 4 680 53
XM_015447203.1 XP_015302689.1 3 - 4 563 53
GenBank mRNA AB179409.1 - 2 - 3 453 53

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0