This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf04388306_m1
See other RPS24 GE Assays ›
Gene Symbol:
RPS24
Gene Name:
ribosomal protein S24
Gene Aliases:
-
Chromosome Location:
Chr.9: 57567494 - 57574676 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf04388306_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPS24
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005565336.2 XP_005565393.1 1 - 2 212 72
XM_005565337.2 XP_005565394.1 1 - 2 212 72
XM_005565338.2 XP_005565395.1 1 - 2 212 72
XM_005565339.2 XP_005565396.1 1 - 2 212 72
XM_005565340.2 XP_005565397.1 1 - 2 212 72
XM_005565341.2 XP_005565398.1 1 - 2 212 72
XM_005565342.2 XP_005565399.1 1 - 2 212 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

ribosomal protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline