This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf04388587_g1
See other ZNF268 GE Assays ›
Gene Symbol:
ZNF268
Gene Name:
zinc finger protein 268
Gene Aliases:
-
Chromosome Location:
Chr.11: 137694366 - 137718763 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf04388587_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZNF268
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005572717.1 XP_005572774.1 4 - 5 552 101
XM_005572718.2 XP_005572775.1 4 - 5 657 101
XM_005572720.2 XP_005572777.1 4 - 5 452 101
XM_005572721.2 XP_005572778.1 4 - 5 564 101
XM_005572722.2 XP_005572779.1 4 - 5 424 101
XM_005572726.2 XP_005572783.1 4 - 5 665 101
XM_015431882.1 XP_015287368.1 4 - 5 620 101
XM_015431883.1 XP_015287369.1 3 - 4 543 101
XM_015431886.1 XP_015287372.1 4 - 5 665 101
XM_015431887.1 XP_015287373.1 4 - 5 459 101

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_005572722.2, XM_015431883.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-6dxlp:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0