This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf04389047_g1
See other BRCA1 GE Assays ›
Gene Symbol:
BRCA1
Gene Name:
BRCA1 DNA repair associated
Gene Aliases:
-
Chromosome Location:
Chr.16: 54819680 - 54913260 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf04389047_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BRCA1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001301366.1 NP_001288295.1 16 - 17 5148 99
XM_005584349.2 XP_005584406.1 17 - 18 5607 99
XM_015437852.1 XP_015293338.1 17 - 18 5601 99
XM_015437853.1 XP_015293339.1 17 - 18 5199 99
XM_015437854.1 XP_015293340.1 17 - 18 5186 99
XM_015437855.1 XP_015293341.1 17 - 18 5594 99
XM_015437856.1 XP_015293342.1 17 - 18 5598 99
XM_015437857.1 XP_015293343.1 17 - 18 5599 99
XM_015437858.1 XP_015293344.1 16 - 17 5523 99
XM_015437859.1 XP_015293345.1 16 - 17 2164 99
GenBank mRNA KM017624.1 - 16 - 17 5148 99

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase protein modifying enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0