Assay Details
| Gene Symbol: LOC102116465 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015455492.1 | XP_015310978.1 | 2 - 3 | 293 | 75 | |
| XM_015455493.1 | XP_015310979.1 | 2 - 3 | 293 | 75 | ||
More Information
Set Membership: |
|

LOC102116465
Genomic Map