This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf04393136_s1
See other TRAPPC2L GE Assays ›
Gene Symbol:
TRAPPC2L
Gene Name:
trafficking protein particle complex 2 like
Gene Aliases:
-
Chromosome Location:
Chr.20: 77304631 - 77310691 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf04393136_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TRAPPC2L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005592789.2 XP_005592846.1 5 1166 67
XM_005592790.2 XP_005592847.1 5 1163 67
XM_015442941.1 XP_015298427.1 7 1384 67
XM_015442942.1 XP_015298428.1 7 1380 67
XM_015442943.1 XP_015298429.1 7 1310 67
XM_015442944.1 XP_015298430.1 7 1307 67
XM_015442945.1 XP_015298431.1 6 1270 67
XM_015442946.1 XP_015298432.1 6 1267 67
XM_015442947.1 XP_015298433.1 6 1230 67
XM_015442948.1 XP_015298434.1 6 1223 67
XM_015442949.1 XP_015298435.1 6 1308 67
XM_015442950.1 XP_015298436.1 6 1790 67
XM_015442951.1 XP_015298437.1 6 1741 67
XM_015442952.1 XP_015298438.1 6 1734 67
XM_015442953.1 XP_015298439.1 6 1258 67
XM_015442954.1 XP_015298440.1 6 1261 67
XM_015442955.1 XP_015298441.1 6 1225 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

membrane traffic protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0