Assay Details
| Gene Symbol: C4H6orf163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284172.1 | NP_001271101.1 | 2 - 3 | 489 | 71 | |
| XM_015448836.1 | XP_015304322.1 | 3 - 4 | 2475 | 71 | ||
| GenBank mRNA | AB070040.1 | - | 2 - 3 | 489 | 71 | |

C4H6orf163
Genomic Map