This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf07267226_s1
See other TMEM25 GE Assays ›
Gene Symbol:
TMEM25
Gene Name:
transmembrane protein 25
Gene Aliases:
-
Chromosome Location:
Chr.14: 114525199 - 114544567 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf07267226_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMEM25
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005579807.2 XP_005579864.1 1 408 60
XM_005579808.2 XP_005579865.1 1 412 60
XM_005579809.2 XP_005579866.1 1 413 60
XM_015435791.1 XP_015291277.1 1 416 60
XM_015435792.1 XP_015291278.1 1 410 60
XM_015435793.1 XP_015291279.1 1 266 60
XM_015435794.1 XP_015291280.1 1 418 60
XM_015435795.1 XP_015291281.1 1 413 60
XM_015435796.1 XP_015291282.1 1 418 60
GenBank mRNA AB050516.1 - 1 318 60

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0