This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf07275627_m1
See other DDX11 GE Assays ›
Gene Symbol:
DDX11
Gene Name:
DEAD/H-box helicase 11
Gene Aliases:
DDX12P
Chromosome Location:
Chr.11: 9745102 - 9779150 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf07275627_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DDX11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005570074.2 XP_005570131.1 13 - 14 2705 98
XM_005570077.1 XP_005570134.1 13 - 14 1661 98
XM_015430346.1 XP_015285832.1 13 - 14 2708 98
XM_015430347.1 XP_015285833.1 13 - 14 2707 98
XM_015430348.1 XP_015285834.1 13 - 14 1541 98
XM_015430349.1 XP_015285835.1 13 - 14 2706 98
XM_015430350.1 XP_015285836.1 13 - 14 2707 98
XM_015430351.1 XP_015285837.1 13 - 14 2707 98
XM_015430352.1 XP_015285838.1 13 - 14 1643 98

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Probe spans exons

Panther Classification:

Molecular Function -

DNA helicase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline