This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00431964_g1
See other ATP5A1 GE Assays ›
Gene Symbol:
Atp5a1
Gene Name:
ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit 1
Gene Aliases:
AI035633, AL022851, AL023067, Atpm, D18Ertd206e, Mom2
Chromosome Location:
Chr.18: 77773768 - 77782869 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00431964_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atp5a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007505.2 NP_031531.1 6 - 7 1082 109
GenBank mRNA AK043976.1 - 6 - 7 850 109
AK076572.1 - 6 - 7 850 109
AK098155.1 - 6 - 7 860 109
AK146797.1 - 6 - 7 853 109
AK150426.1 - 6 - 7 853 109
AK150843.1 - 6 - 7 850 109
AK151004.1 - 6 - 7 850 109
AK151128.1 - 6 - 7 859 109
AK151224.1 - 6 - 7 857 109
AK151920.1 - 6 - 7 854 109
AK152054.1 - 6 - 7 861 109
AK152890.1 - 6 - 7 853 109
AK153167.1 - 6 - 7 854 109
AK153282.1 - 6 - 7 850 109
AK157529.1 - 4 - 5 631 109
AK159491.1 - 6 - 7 850 109
AK159540.1 - 6 - 7 850 109
AK159758.1 - 6 - 7 853 109
AK160043.1 - 6 - 7 860 109
AK164110.1 - 6 - 7 853 109
AK164193.1 - 4 - 5 556 109
AK166709.1 - 6 - 7 853 109
AK166812.1 - 6 - 7 866 109
AK167159.1 - 6 - 7 850 109
AK167863.1 - 6 - 7 830 109
AK168198.1 - 6 - 7 860 109
AK168369.1 - 6 - 7 852 109
AK168879.1 - 6 - 7 850 109
AK168890.1 - 6 - 7 850 109
AK168932.1 - 6 - 7 850 109
AK169080.1 - 6 - 7 853 109
AK169084.1 - 6 - 7 853 109
AK169105.1 - 6 - 7 850 109
AK169142.1 - 6 - 7 895 109
AK169300.1 - 6 - 7 850 109
AK169308.1 - 6 - 7 855 109
AK169414.1 - 6 - 7 853 109
BC014854.1 - 6 - 7 837 4500893 109
BC020417.1 - 6 - 7 837 109
BC055892.1 - 6 - 7 1287 109
L01062.1 - 6 - 7 837 109

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800000833.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atp5a1

Panther Classification:

Molecular Function -

ATP synthase

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of endothelial cell proliferation
lipid metabolic process
ATP biosynthetic process
transport
ion transport
embryo development
ATP synthesis coupled proton transport
ATP hydrolysis coupled proton transport
proton transport
ATP metabolic process
hydrogen ion transmembrane transport
nucleotide binding
ATP binding
hydrolase activity, acting on acid anhydrides, catalyzing transmembrane movement of substances
ATPase activity
MHC class I protein binding
ADP binding
poly(A) RNA binding
proton-transporting ATP synthase activity, rotational mechanism
proton-transporting ATPase activity, rotational mechanism

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800006087.mm.1 NM_007505.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800006087.mm.1 NM_007505.2 Refseq
Affymetrix Mouse Clariom S Array PSR1800006087.mm.1 TC1800000833.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0