This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00431967_g1
See other ATP5A1 GE Assays ›
Gene Symbol:
Atp5a1
Gene Name:
ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit 1
Gene Aliases:
AI035633, AL022851, AL023067, Atpm, D18Ertd206e, Mom2
Chromosome Location:
Chr.18: 77773768 - 77782869 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00431967_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atp5a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007505.2 NP_031531.1 9 - 10 1565 62
GenBank mRNA AK043976.1 - 9 - 10 1333 62
AK076572.1 - 9 - 10 1333 62
AK098155.1 - 9 - 10 1343 62
AK150426.1 - 9 - 10 1336 62
AK150843.1 - 9 - 10 1333 62
AK151004.1 - 9 - 10 1333 62
AK151128.1 - 9 - 10 1342 62
AK151224.1 - 9 - 10 1340 62
AK151920.1 - 9 - 10 1337 62
AK152054.1 - 9 - 10 1344 62
AK152890.1 - 9 - 10 1336 62
AK153167.1 - 9 - 10 1337 62
AK153282.1 - 9 - 10 1333 62
AK159491.1 - 9 - 10 1333 62
AK159540.1 - 9 - 10 1333 62
AK159758.1 - 9 - 10 1336 62
AK160043.1 - 9 - 10 1343 62
AK164110.1 - 9 - 10 1336 62
AK164193.1 - 7 - 8 1039 62
AK166709.1 - 9 - 10 1336 62
AK166812.1 - 9 - 10 1349 62
AK167159.1 - 9 - 10 1333 62
AK168198.1 - 9 - 10 1343 62
AK168369.1 - 9 - 10 1335 62
AK168617.1 - 9 - 10 1333 62
AK168879.1 - 9 - 10 1333 62
AK168890.1 - 9 - 10 1333 62
AK168932.1 - 9 - 10 1333 62
AK169080.1 - 9 - 10 1336 62
AK169084.1 - 9 - 10 1336 62
AK169105.1 - 9 - 10 1333 62
AK169142.1 - 9 - 10 1378 62
AK169300.1 - 9 - 10 1333 62
AK169308.1 - 9 - 10 1338 62
AK169414.1 - 9 - 10 1336 62
BC014854.1 - 9 - 10 1320 4500893 62
BC020417.1 - 9 - 10 1320 62
BC055892.1 - 9 - 10 1770 62
L01062.1 - 9 - 10 1320 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800000833.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atp5a1

Panther Classification:

Molecular Function -

ATP synthase

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of endothelial cell proliferation
lipid metabolic process
ATP biosynthetic process
transport
ion transport
embryo development
ATP synthesis coupled proton transport
ATP hydrolysis coupled proton transport
proton transport
ATP metabolic process
hydrogen ion transmembrane transport
nucleotide binding
ATP binding
hydrolase activity, acting on acid anhydrides, catalyzing transmembrane movement of substances
ATPase activity
MHC class I protein binding
ADP binding
poly(A) RNA binding
proton-transporting ATP synthase activity, rotational mechanism
proton-transporting ATPase activity, rotational mechanism

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800006092.mm.1 NM_007505.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800006092.mm.1 NM_007505.2 Refseq
Affymetrix Mouse Clariom S Array PSR1800006092.mm.1 TC1800000833.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0