This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00432019_m1
See other SLC7A1 GE Assays ›
Gene Symbol:
Slc7a1
Gene Name:
solute carrier family 7 (cationic amino acid transporter, y+ system), member 1
Gene Aliases:
4831426K01Rik, AI447493, Atrc-1, Atrc1, Cat1, Rec-1, Rev-1, mCAT-1
Chromosome Location:
Chr.5: 148327410 - 148399904 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00432019_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc7a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007513.4 NP_031539.3 1 - 2 176 73
GenBank mRNA AK139342.1 - 1 - 2 152 73
AK147991.1 - 1 - 2 85 73
BC068234.1 - 1 - 2 68 73
CJ098794.1 - 1 - 2 177 73
M26687.1 - 1 - 2 94 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500003637.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK139342.1,AK147991.1,M26687.1,NM_007513.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc7a1

Panther Classification:

Molecular Function -

amino acid transporter

Gene Ontology Categories:

Function(s) Process(es)

amino acid transmembrane transport
transport
amino acid transport
arginine transport
regulation of TOR signaling
L-arginine import across plasma membrane
L-ornithine transmembrane transport
L-lysine transmembrane transport
arginine transmembrane transport
L-ornithine transmembrane transporter activity
amino acid transmembrane transporter activity
arginine transmembrane transporter activity
L-lysine transmembrane transporter activity
antiporter activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500033386.mm.1 TC0500003637.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500033386.mm.1 NM_007513.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500033386.mm.1 NM_007513.4 Refseq
Affymetrix Mouse Clariom S Array PSR0500033387.mm.1 TC0500003637.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500033387.mm.1 NM_007513.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500033387.mm.1 NM_007513.4 Refseq
Affymetrix Mouse Clariom S Array PSR0500033393.mm.1 TC0500003637.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500033393.mm.1 NM_007513.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500033393.mm.1 NM_007513.4 Refseq
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 835595 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0