This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00432290_g1
See other CAMK2B GE Assays ›
Gene Symbol:
Camk2b
Gene Name:
calcium/calmodulin-dependent protein kinase II, beta
Gene Aliases:
-
Chromosome Location:
Chr.11: 5969642 - 6065748 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00432290_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Camk2b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001174053.1 NP_001167524.1 9 - 10 903 123
NM_001174054.1 NP_001167525.1 10 - 11 936 123
NM_007595.5 NP_031621.3 9 - 10 903 123
GenBank mRNA AK046454.1 - 9 - 10 904 123
AK158794.1 - 6 - 7 664 123
AK163698.1 - 9 - 10 720 123
BC024803.1 - 10 - 11 752 123
BC080273.1 - 9 - 10 705 6851995 123
X63615.1 - 9 - 10 718 123

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100002158.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Camk2b

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
inhibitory G-protein coupled receptor phosphorylation
protein phosphorylation
calcium ion transport
nervous system development
positive regulation of neuron projection development
phosphorylation
peptidyl-serine phosphorylation
cell differentiation
regulation of synaptic transmission, cholinergic
positive regulation of phospholipase A2 activity
response to cadmium ion
protein autophosphorylation
regulation of long-term neuronal synaptic plasticity
cell projection morphogenesis
neuromuscular process controlling balance
long-term synaptic potentiation
activation of meiosis involved in egg activation
positive regulation of dendritic spine morphogenesis
positive regulation of synapse maturation
positive regulation of apoptotic signaling pathway
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
calmodulin-dependent protein kinase activity
protein binding
calmodulin binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
protein homodimerization activity
phospholipase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0