This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00432832_g1
See other DLD GE Assays ›
Gene Symbol:
Dld
Gene Name:
dihydrolipoamide dehydrogenase
Gene Aliases:
-
Chromosome Location:
Chr.12: 31331562 - 31351471 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00432832_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dld
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007861.5 NP_031887.2 6 - 7 570 113
GenBank mRNA AK117104.1 - 6 - 7 514 113
AK135807.1 - 6 - 7 513 113
AK167885.1 - 6 - 7 514 113
AK168157.1 - 6 - 7 535 113
AK168875.1 - 6 - 7 455 113
BC003368.1 - 6 - 7 497 3589344 113
U73445.1 - 6 - 7 547 113

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dld

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

acetyl-CoA biosynthetic process from pyruvate
2-oxoglutarate metabolic process
mitochondrial electron transport, NADH to ubiquinone
proteolysis
gastrulation
lipoate metabolic process
regulation of membrane potential
cell redox homeostasis
sperm capacitation
dihydrolipoamide metabolic process
oxidation-reduction process
dihydrolipoyl dehydrogenase activity
oxidoreductase activity
oxidoreductase activity, acting on a sulfur group of donors, NAD(P) as acceptor
lipoamide binding
flavin adenine dinucleotide binding
NAD binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-6dxlp:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0