This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00433550_g1
See other GCGR GE Assays ›
Gene Symbol:
Gcgr
Gene Name:
glucagon receptor
Gene Aliases:
GR
Chromosome Location:
Chr.11: 120529190 - 120538987 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00433550_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gcgr
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008101.2 NP_032127.2 7 - 8 846 75
GenBank mRNA AK144393.1 - 7 - 8 793 75
BC012718.1 - 6 - 7 921 75
BC031885.1 - 7 - 8 796 5137340 75
BC057988.1 - 7 - 8 777 5100574 75
L38613.1 - 6 - 7 756 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100002032.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gcgr

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

exocytosis
signal transduction
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
adenylate cyclase-activating G-protein coupled receptor signaling pathway
hormone-mediated signaling pathway
glucose homeostasis
response to starvation
regulation of glycogen metabolic process
cellular response to glucagon stimulus
signal transducer activity
transmembrane signaling receptor activity
G-protein coupled receptor activity
glucagon receptor activity
peptide hormone binding

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28185093 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28185092 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28185090 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28185091 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28185094 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2056995 L38613.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2056995 BC057988.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2056995 AK144393.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2056995 BC012718.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2056995 BC031885.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 716347 AK017485.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 716347 AK161639.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 716347 BC048919.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 716347 XM_001004179.1 refseq_mrna_mouse_XM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 716347 XM_489285.3 refseq_mrna_mouse_XM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 456449 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 924378 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 91120 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 716561 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 534009 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 136821 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P124993 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2056995 L38613.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2056995 BC057988.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2056995 AK144393.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2056995 BC012718.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2056995 BC031885.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P211403 L38613.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185110 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185086 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185083 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185076 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185104 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185106 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185073 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185092 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185097 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185091 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185075 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185107 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185103 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185087 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185099 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185085 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185067 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185113 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185080 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185089 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185112 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185101 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185090 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185108 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185072 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185084 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185071 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185079 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28185094 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P124993 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02446608 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07516604 - -
Whole Mouse Genome G4122A A_51_P211403 L38613.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113121 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P124993 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09520652 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09520660 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09240837 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09520647 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09520666 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28185093 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28185092 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28185094 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28185091 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28185090 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185061 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185103 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185083 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185179 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185090 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185121 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185127 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185205 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185075 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185142 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185163 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185091 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185076 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185181 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185208 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185151 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185197 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185063 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185183 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185212 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185177 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185137 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185226 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185195 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185080 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185215 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185170 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185209 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185052 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185056 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185140 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185216 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185207 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185223 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185162 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185089 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185158 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185217 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185087 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185159 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185186 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185092 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185198 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185101 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185160 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185214 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185097 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185219 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185122 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185153 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185221 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185107 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185099 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185054 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185169 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185220 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185204 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185200 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185202 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185133 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185189 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185143 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185085 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185173 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185117 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185145 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185130 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185115 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185106 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185120 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185135 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185182 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185218 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185104 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185113 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185094 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185050 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185157 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185167 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185124 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185155 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185068 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185224 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185132 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185191 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185156 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185066 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185185 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185147 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185150 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185071 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28185108 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185163 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185153 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185097 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185110 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185085 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185132 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185137 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185079 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185064 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185155 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09616995 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185080 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185094 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09270125 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185130 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185142 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185151 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185107 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185073 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09270168 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185104 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185135 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185124 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185063 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185083 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185114 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185065 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09273579 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185167 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185133 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185156 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185068 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185127 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185090 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09270172 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09270157 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09270155 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185075 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185076 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185169 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185120 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185174 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09270158 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185101 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09240841 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185121 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185074 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185087 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185066 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185173 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185113 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185108 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185140 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09240827 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185143 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09531690 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185089 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09156733 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185106 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185147 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185118 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185145 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28185071 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09191797 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10383422 - -
Affymetrix Mouse Gene 1.0 ST Array 10383420 - -
Affymetrix Mouse Gene 1.0 ST Array 10383411 - -
Affymetrix Mouse Gene 1.0 ST Array 10383410 - -
Affymetrix Mouse Gene 1.0 ST Array 10383416 - -
Affymetrix Mouse Gene 1.0 ST Array 10383412 - -
Affymetrix Mouse Gene 1.0 ST Array 10383421 - -
Affymetrix Mouse Gene 1.0 ST Array 10383418 - -
Affymetrix Mouse Gene 1.0 ST Array 10383415 - -
Affymetrix Mouse Gene 1.0 ST Array 10383414 - -
Affymetrix Mouse Gene 1.0 ST Array 10383419 - -
Affymetrix Mouse Gene 1.0 ST Array 10383417 - -
Affymetrix Mouse Gene 1.0 ST Array 10383413 - -
Affymetrix HT MG-430 PM Array Plate 1450127_PM_a_at - -
Affymetrix Mouse Gene 1.1 ST Array 10383409 - -
Mouse Genome 430 2.0 Array 1450127_a_at - -
Mouse Genome 430 2.0 Array 1450127_PM_a_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 14527_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 14527_PM_at - -
Affymetrix Mouse Clariom S Array PSR1100019588.mm.1 NM_008101.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100019588.mm.1 NM_008101.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100019588.mm.1 TC1100002032.mm.1 MTA TC
Mouse Expression Set 430 1450127_PM_a_at - -
Mouse Expression Set 430 1450127_a_at - -
Murine Genome U74v2 Set 103498_at - -
Murine Genome U74v2 Set 103498_PM_at - -
Mouse Genome 430A 2.0 Array 1450127_PM_a_at - -
Mouse Genome 430A 2.0 Array 1450127_a_at - -
Affymetrix HT Mouse Genome 430A Array 1450127_a_at - -
Affymetrix HT Mouse Genome 430A Array 1450127_PM_a_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 6620497 - -
Illumina mouse-6 v1.0 expression beadchip 6620497 - -
Illumina mouseRef-8 v1.0 expression beadchip 0006620497 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2623764 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2623764 - -
mouse-6 v1.1 (Illumina) scl40629.9_47-S - -
Illumina Sentrix mouse-6.v1 bead array 6620497 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl40629.9_47-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 6620497 - -
MouseRef-8 v2 Expression BeadChip ILMN_2874278 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2874278 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-cj8gb:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline