Assay Details
| Gene Symbol: Gpld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008156.2 | NP_032182.2 | 9 - 10 | 969 | 64 | |
| GenBank mRNA | AF050666.1 | - | 9 - 10 | 911 | 64 | |
| AK004957.2 | - | 8 - 9 | 673 | 64 | ||
| AY081194.1 | - | 8 - 9 | 696 | 64 | ||
| AY333960.1 | - | 8 - 9 | 653 | 64 | ||
| BC019146.1 | - | 8 - 9 | 682 | 5052822 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpld1
Genomic Map