This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00433978_m1
See other LIPC GE Assays ›
Gene Symbol:
Lipc
Gene Name:
lipase, hepatic
Gene Aliases:
AI256194, HL, Hpl
Chromosome Location:
Chr.9: 70798128 - 70952221 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00433978_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lipc
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008280.2 NP_032306.2 6 - 7 1309 83
GenBank mRNA AK158285.1 - 6 - 7 1160 83
AK158360.1 - 6 - 7 1160 83
AY228765.1 - 6 - 7 1061 83
BC021841.1 - 6 - 7 1309 5124139 83
BC094050.1 - 6 - 7 1096 5135403 83
X58426.1 - 6 - 7 1080 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900002601.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Lipc

Panther Classification:

Molecular Function -

lipase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
fatty acid metabolic process
fatty acid biosynthetic process
triglyceride metabolic process
phosphatidylserine metabolic process
cholesterol metabolic process
heparan sulfate proteoglycan biosynthetic process
lipid catabolic process
triglyceride catabolic process
cholesterol transport
chylomicron remodeling
very-low-density lipoprotein particle remodeling
low-density lipoprotein particle remodeling
high-density lipoprotein particle remodeling
chylomicron remnant clearance
low-density lipoprotein particle clearance
cholesterol homeostasis
phosphatidylethanolamine metabolic process
neutral lipid catabolic process
phosphatidylcholine metabolic process
phosphatidic acid metabolic process
glycerophospholipid catabolic process
protein oligomerization
triglyceride homeostasis
regulation of plasma lipoprotein particle levels
lysophospholipase activity
triglyceride lipase activity
protein binding
heparin binding
lipid binding
phosphatidylcholine 1-acylhydrolase activity
lipase activity
transferase activity, transferring acyl groups
hydrolase activity
low-density lipoprotein particle binding
apolipoprotein binding
chylomicron binding
heparan sulfate proteoglycan binding
acylglycerol lipase activity
carboxylic ester hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900021628.mm.1 NM_008280.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900021628.mm.1 NM_008280.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900021628.mm.1 TC0900002601.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-z47m5:80/100.66.77.18:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0