This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00434235_m1
See other IL1R1 GE Assays ›
Gene Symbol:
Il1r1
Gene Name:
interleukin 1 receptor, type I
Gene Aliases:
CD121a, CD121b, IL-1R1, IL-iR, Il1r-1
Chromosome Location:
Chr.1: 40225014 - 40316201 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00434235_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Il1r1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001123382.1 NP_001116854.1 4 - 5 787 110
NM_008362.2 NP_032388.1 4 - 5 718 110
GenBank mRNA AK048550.1 - 4 - 5 705 110
AK054137.1 - 4 - 5 788 110
BC109135.2 - 4 - 5 624 40039321 110
M20658.1 - 4 - 5 718 110

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Il1r1

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
response to radiation
heat acclimation
cytokine-mediated signaling pathway
regulation of inflammatory response
interleukin-1-mediated signaling pathway
response to interleukin-1
protease binding
signal transducer activity
interleukin-1 receptor activity
interleukin-1, Type I, activating receptor activity
platelet-derived growth factor receptor binding
protein binding
ionotropic glutamate receptor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0