Assay Details
| Gene Symbol: Il7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008371.4 | NP_032397.1 | 1 - 2 | 251 | 101 | |
| GenBank mRNA | AK041307.1 | - | 1 - 2 | 252 | 101 | |
| BC110553.2 | - | 1 - 2 | 160 | 40047584 | 101 | |
| X07962.1 | - | 1 - 2 | 551 | 101 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK041307.1,BC110553.2,NM_008371.4,X07962.1 transcripts.
Set Membership: |
|

Il7
Genomic Map