This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00434767_m1
See other LPL GE Assays ›
Gene Symbol:
Lpl
Gene Name:
lipoprotein lipase
Gene Aliases:
-
Chromosome Location:
Chr.8: 68880555 - 68906932 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00434767_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lpl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008509.2 NP_032535.2 5 - 6 980 92
GenBank mRNA AK002645.1 - 5 - 6 906 92
AK017272.1 - 978 92
AK045064.1 - 5 - 6 869 92
AK079446.1 - 5 - 6 869 92
AK086023.1 - 5 - 6 801 92
AK150328.1 - 5 - 6 981 92
AK150355.1 - 5 - 6 981 92
AK150375.1 - 5 - 6 979 92
AK150457.1 - 5 - 6 876 92
AK150488.1 - 5 - 6 981 92
AK150505.1 - 5 - 6 981 92
AK150532.1 - 5 - 6 981 92
AK150577.1 - 5 - 6 981 92
AK150585.1 - 5 - 6 981 92
AK150593.1 - 5 - 6 981 92
AK150646.1 - 5 - 6 978 92
AK150672.1 - 5 - 6 899 92
AK150691.1 - 5 - 6 981 92
AK150984.1 - 5 - 6 981 92
AK150990.1 - 5 - 6 981 92
AK151006.1 - 5 - 6 981 92
AK151038.1 - 5 - 6 879 92
AK151049.1 - 5 - 6 981 92
AK151093.1 - 5 - 6 981 92
AK151099.1 - 5 - 6 981 92
AK151105.1 - 5 - 6 981 92
AK151112.1 - 5 - 6 982 92
AK151195.1 - 5 - 6 879 92
AK151216.1 - 5 - 6 982 92
AK151369.1 - 5 - 6 899 92
AK151434.1 - 5 - 6 899 92
AK151521.1 - 5 - 6 899 92
AK151540.1 - 5 - 6 869 92
AK151563.1 - 5 - 6 981 92
AK151630.1 - 5 - 6 879 92
AK151679.1 - 5 - 6 981 92
AK151727.1 - 5 - 6 981 92
AK151772.1 - 5 - 6 981 92
AK151801.1 - 5 - 6 869 92
AK151828.1 - 5 - 6 869 92
AK151866.1 - 5 - 6 981 92
AK151870.1 - 5 - 6 981 92
AK151872.1 - 5 - 6 981 92
AK151893.1 - 5 - 6 869 92
AK152014.1 - 5 - 6 981 92
AK152035.1 - 5 - 6 981 92
AK152049.1 - 5 - 6 981 92
AK152053.1 - 5 - 6 869 92
AK152079.1 - 5 - 6 981 92
AK152238.1 - 5 - 6 982 92
AK152251.1 - 5 - 6 979 92
AK152350.1 - 5 - 6 981 92
AK152390.1 - 5 - 6 820 92
AK152481.1 - 5 - 6 982 92
AK152657.1 - 5 - 6 981 92
AK152706.1 - 5 - 6 982 92
AK152733.1 - 5 - 6 879 92
AK152869.1 - 5 - 6 981 92
AK152956.1 - 5 - 6 982 92
AK153006.1 - 5 - 6 92
AK153148.1 - 5 - 6 981 92
AK153242.1 - 5 - 6 982 92
AK153425.1 - 5 - 6 981 92
AK153428.1 - 5 - 6 981 92
AK159268.1 - 5 - 6 981 92
AK170486.1 - 5 - 6 981 92
BC003305.1 - 5 - 6 969 3584788 92
BC158040.1 - 5 - 6 814 9088156 92
CT010344.1 - 5 - 6 782 92
J03302.1 - 5 - 6 758 92

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800000707.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Lpl

Panther Classification:

Molecular Function -

lipase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
fatty acid biosynthetic process
positive regulation of macrophage derived foam cell differentiation
positive regulation of cholesterol storage
positive regulation of sequestering of triglyceride
lipid catabolic process
triglyceride biosynthetic process
triglyceride catabolic process
very-low-density lipoprotein particle remodeling
triglyceride homeostasis
lipoprotein lipase activity
triglyceride lipase activity
receptor binding
protein binding
heparin binding
hydrolase activity
triglyceride binding
apolipoprotein binding
carboxylic ester hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800004750.mm.1 NM_008509.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800004750.mm.1 NM_008509.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800004750.mm.1 TC0800000707.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800004751.mm.1 NM_008509.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800004751.mm.1 NM_008509.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800004751.mm.1 TC0800000707.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vh84l:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0