This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00435009_m1
See other MTTP GE Assays ›
Gene Symbol:
Mttp
Gene Name:
microsomal triglyceride transfer protein
Gene Aliases:
1810043K16Rik, MTP
Chromosome Location:
Chr.3: 138089853 - 138143388 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00435009_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mttp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163457.1 NP_001156929.1 12 - 13 1727 72
NM_008642.2 NP_032668.2 10 - 11 1417 72
GenBank mRNA AK136278.1 - 12 - 13 1727 72
AK146553.1 - 10 - 11 1418 72
BC012686.1 - 10 - 11 1377 4163901 72
EU553486.1 - 10 - 11 1385 72
L47970.1 - 10 - 11 1364 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300003016.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mttp

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

protein lipidation
lipid metabolic process
triglyceride metabolic process
transport
lipid transport
steroid metabolic process
cholesterol metabolic process
lipoprotein metabolic process
cholesterol homeostasis
lipoprotein transport
lipid transporter activity
protein binding
lipid binding
apolipoprotein binding
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300023787.mm.1 NM_001163457.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300023787.mm.1 NM_001163457.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300023787.mm.1 NM_008642.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300023787.mm.1 NM_008642.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300023787.mm.1 TC0300003016.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0