This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00435165_m1
See other NNT GE Assays ›
Gene Symbol:
Nnt
Gene Name:
nicotinamide nucleotide transhydrogenase
Gene Aliases:
4930423F13Rik, AI323702, BB168308
Chromosome Location:
Chr.13: 119335527 - 119409138 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00435165_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nnt
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008710.3 NP_032736.2 2621 64
GenBank mRNA AK033282.1 - 11 - 12 1809 64
AK040460.1 - 9 - 10 1487 64
AK050478.1 - 10 - 11 2526 64
AK052234.1 - 11 - 12 1853 64
AK054332.1 - 11 - 12 1854 64
AK083413.1 - 11 - 12 1852 64
AK084546.1 - 11 - 12 1851 64
AK084604.1 - 11 - 12 1851 64
AK085903.1 - 9 - 10 1512 64
AK088044.1 - 11 - 12 2603 64
AK090115.1 - 2624 64
AK129006.1 - 11 - 12 2598 64
AK159694.1 - 12 - 13 2607 64
AK168736.1 - 10 - 11 2404 64
BC008518.1 - 11 - 12 2584 3601702 64
Z49204.1 - 10 - 11 2528 64

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nnt

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

proton transport
cell redox homeostasis
oxidation-reduction process
NAD(P)+ transhydrogenase activity
NAD(P)+ transhydrogenase (AB-specific) activity
oxidoreductase activity
NADP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline