This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00435351_m1
See other SLC11A2 GE Assays ›
Gene Symbol:
Slc11a2
Gene Name:
solute carrier family 11 (proton-coupled divalent metal ion transporters), member 2
Gene Aliases:
DCT1, DMT1, Nramp2, mk, van
Chromosome Location:
Chr.15: 100387898 - 100424209 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00435351_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc11a2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001146161.1 NP_001139633.1 2 - 3 169 84
NM_008732.2 NP_032758.2 2 - 3 169 84
GenBank mRNA AF029758.1 - 2 - 3 104 84
AK049856.1 - 2 - 3 117 84
AK083478.1 - 2 - 3 115 84
BC019137.1 - 2 - 3 143 5040976 84
CJ057283.1 - 2 - 3 170 84
L33415.1 - 2 - 3 97 84

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF029758.1, AK049856.1, AK083478.1, BC019137.1, L33415.1, NM_001146161.1, NM_008732.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc11a2

Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

transition metal ion transport
porphyrin-containing compound metabolic process
porphyrin-containing compound biosynthetic process
heme biosynthetic process
transport
ion transport
cobalt ion transport
copper ion transport
iron ion transport
manganese ion transport
cellular copper ion homeostasis
activation of cysteine-type endopeptidase activity involved in apoptotic process
learning or memory
response to manganese ion
nickel cation transport
vanadium ion transport
ferrous iron transport
lead ion transport
proton transport
cellular response to oxidative stress
iron ion transmembrane transport
copper ion transmembrane transport
nickel cation transmembrane transport
dendrite morphogenesis
erythrocyte development
iron ion homeostasis
multicellular organismal iron ion homeostasis
cadmium ion transmembrane transport
ferrous iron import
manganese ion transmembrane transport
zinc II ion transmembrane transport
cation transmembrane transport
hydrogen ion transmembrane transport
copper ion import into cell
ferrous iron transmembrane transport
transporter activity
copper ion transmembrane transporter activity
iron ion transmembrane transporter activity
manganese ion transmembrane transporter activity
iron ion binding
copper ion binding
zinc ion binding
hydrogen ion transmembrane transporter activity
cadmium ion transmembrane transporter activity
cobalt ion transmembrane transporter activity
ferrous iron transmembrane transporter activity
lead ion transmembrane transporter activity
nickel cation transmembrane transporter activity
vanadium ion transmembrane transporter activity
solute:proton symporter activity
ferrous iron uptake transmembrane transporter activity
nickel cation binding
inorganic cation transmembrane transporter activity
manganese ion binding
cadmium ion binding
transition metal ion transmembrane transporter activity
cobalt ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0