Assay Details
| Gene Symbol: Slc11a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146161.1 | NP_001139633.1 | 13 - 14 | 1482 | 94 | |
| NM_008732.2 | NP_032758.2 | 13 - 14 | 1482 | 94 | ||
| GenBank mRNA | AF029758.1 | - | 13 - 14 | 1417 | 94 | |
| AK049856.1 | - | 13 - 14 | 1430 | 94 | ||
| AK083478.1 | - | 13 - 14 | 1428 | 94 | ||
| AK148276.1 | - | 13 - 14 | 1430 | 94 | ||
| BC019137.1 | - | 13 - 14 | 1456 | 5040976 | 94 | |
| L33415.1 | - | 13 - 14 | 1410 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc11a2
Genomic Map