This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00435451_m1
See other TNFRSF11B GE Assays ›
Gene Symbol:
Tnfrsf11b
Gene Name:
tumor necrosis factor receptor superfamily, member 11b (osteoprotegerin)
Gene Aliases:
OCIF, Opg, TR1
Chromosome Location:
Chr.15: 54250619 - 54278484 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00435451_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tnfrsf11b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008764.3 NP_032790.3 1 - 2 269 74
GenBank mRNA AB013898.1 - 1 - 2 269 74
AK140244.1 - 1 - 2 92 74
AK146105.1 - 1 - 2 212 74
BC049782.1 - 1 - 2 86 6506550 74
U94331.1 - 1 - 2 122 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500001528.mm.1

Additional Information:

This assay is in the 5'UTR region of the AB013898.1,AK140244.1,AK146105.1,BC049782.1,NM_008764.3,U94331.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tnfrsf11b

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s)

apoptotic process
signal transduction
extracellular matrix organization
negative regulation of odontogenesis of dentin-containing tooth
negative regulation of bone resorption

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 513219 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-t8jrk:80/100.66.77.131:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0