Assay Details
| Gene Symbol: Prkci | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008857.3 | NP_032883.2 | 14 - 15 | 1620 | 67 | |
| GenBank mRNA | AK144979.1 | - | 14 - 15 | 1620 | 67 | |
| AK147710.1 | - | 14 - 15 | 1592 | 67 | ||
| AK167411.1 | - | 14 - 15 | 1589 | 67 | ||
| AK220517.1 | - | 14 - 15 | 1644 | 67 | ||
| BC021630.1 | - | 14 - 15 | 1491 | 3980628 | 67 | |
| D28577.1 | - | 14 - 15 | 1415 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prkci
Genomic Map