This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00437052_m1
See other TIAL1 GE Assays ›
Gene Symbol:
Tial1
Gene Name:
Tia1 cytotoxic granule-associated RNA binding protein-like 1
Gene Aliases:
5330433G13Rik, AL033329, TIAR, mTIAR
Chromosome Location:
Chr.7: 128439772 - 128461668 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00437052_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tial1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009383.2 NP_033409.1 5 - 6 784 67
GenBank mRNA AK012085.1 - 5 - 6 428 67
AK132677.1 - 6 - 7 954 67
BC010496.1 - 6 - 7 518 3497971 67
BC068194.1 - 1 - 2 60 67
U55861.1 - 5 - 6 560 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700004366.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tial1

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
germ cell development
positive regulation of cell proliferation
stem cell division
nucleotide binding
nucleic acid binding
RNA binding
protein binding
AU-rich element binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700036685.mm.1 NM_009383.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700036685.mm.1 NM_009383.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700036685.mm.1 TC0700004366.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0