This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00437138_g1
See other TNFRSF18 GE Assays ›
Gene Symbol:
Tnfrsf18
Gene Name:
tumor necrosis factor receptor superfamily, member 18
Gene Aliases:
AITR, Gitr
Chromosome Location:
Chr.4: 156026342 - 156028891 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00437138_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tnfrsf18
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009400.2 NP_033426.1 3 - 4 407 111
GenBank mRNA AF229432.1 - 3 - 4 407 111
AF229433.1 - 3 - 4 407 111
AK020762.1 - 3 - 4 428 111
AK081878.1 - 3 - 4 396 111
AY157832.1 - 3 - 4 407 111
AY157833.1 - 3 - 4 407 111
U82534.1 - 3 - 4 407 111

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400002104.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tnfrsf18

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of leukocyte migration
apoptotic process
positive regulation of tyrosine phosphorylation of Stat1 protein
positive regulation of cell adhesion
protein binding

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23441977 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23441975 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23441978 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23441979 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23441976 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2025490 U82534.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2025490 AF229433.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2025490 AF229434.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2025490 NM_009400.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2025490 NM_021985.2 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 903677 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 294689 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 166814 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P110329 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2025490 U82534.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2025490 AF229433.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2025490 AF229434.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2025490 NM_009400.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2025490 NM_021985.2 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P469401 U82534.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P469401 AF229433.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P469401 AF229434.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P469401 AK020762.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P469401 NM_009400.2 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441910 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441925 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441920 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441900 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441903 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441929 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441912 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441931 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441922 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441897 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441917 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441926 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441898 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441916 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441927 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441902 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441901 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441915 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441935 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441909 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441907 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441934 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441905 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441933 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441919 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441924 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441913 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441930 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441932 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441894 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441923 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P110329 - -
Whole Mouse Genome G4122A A_51_P469401 U82534.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P469401 AF229433.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P469401 AF229434.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P469401 AK020762.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P469401 NM_009400.2 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120726 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P110329 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04089307 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04089282 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04089283 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04089299 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04089289 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23441979 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23441977 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23441975 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23441976 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23441978 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441941 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441895 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441946 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441916 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441936 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441940 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441949 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441939 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441922 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441893 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441894 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441913 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441932 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441905 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441935 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441917 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441924 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441923 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441920 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441891 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441929 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441948 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441903 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441900 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441927 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441933 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441909 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441934 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441945 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441898 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441926 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23441943 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04089303 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23441888 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23441894 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23441948 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23441905 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03202353 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03961264 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23441920 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03603827 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23441925 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03398887 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23441893 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03854214 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23441898 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23441929 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23441931 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04089304 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23441912 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23441907 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23441940 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03398886 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04089301 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23441924 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23441937 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23441917 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03992745 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03854211 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23441933 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23441897 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23441913 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23441934 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23441900 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03768328 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23441916 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23441935 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23441936 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04089298 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23441949 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23441939 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23441909 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23441895 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10511292 - -
Affymetrix Mouse Gene 1.0 ST Array 10511291 - -
Affymetrix Mouse Gene 1.0 ST Array 10511295 - -
Affymetrix Mouse Gene 1.0 ST Array 10511294 - -
Affymetrix Mouse Gene 1.0 ST Array 10511296 - -
Affymetrix Mouse Gene 1.0 ST Array 10511297 - -
Affymetrix Mouse Gene 1.0 ST Array 10511293 - -
Affymetrix HT MG-430 PM Array Plate 1422303_PM_a_at - -
Affymetrix Mouse Gene 1.1 ST Array 10511290 - -
Mouse Genome 430 2.0 Array 1422303_PM_a_at - -
Mouse Genome 430 2.0 Array 1422303_a_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 21936_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 21936_PM_at - -
Affymetrix Mouse Clariom S Array PSR0400017747.mm.1 NM_009400.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400017747.mm.1 NM_009400.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400017747.mm.1 NM_021985.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400017747.mm.1 NM_021985.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400017747.mm.1 TC0400002104.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400017750.mm.1 NM_009400.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400017750.mm.1 NM_009400.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400017750.mm.1 NM_021985.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400017750.mm.1 NM_021985.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400017750.mm.1 TC0400002104.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400017748.mm.1 NM_009400.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400017748.mm.1 NM_009400.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400017748.mm.1 NM_021985.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400017748.mm.1 NM_021985.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400017748.mm.1 TC0400002104.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400017743.mm.1 NM_009400.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400017743.mm.1 NM_009400.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400017743.mm.1 TC0400002104.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400017745.mm.1 NM_009400.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400017745.mm.1 NM_009400.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400017745.mm.1 TC0400002104.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400017749.mm.1 NM_009400.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400017749.mm.1 NM_009400.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400017749.mm.1 NM_021985.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400017749.mm.1 NM_021985.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400017749.mm.1 TC0400002104.mm.1 MTA TC
Mouse Expression Set 430 1422303_PM_a_at - -
Mouse Expression Set 430 1422303_a_at - -
Murine Genome U74v2 Set 104662_PM_at - -
Murine Genome U74v2 Set 104662_at - -
Mouse Genome 430A 2.0 Array 1422303_a_at - -
Mouse Genome 430A 2.0 Array 1422303_PM_a_at - -
Affymetrix HT Mouse Genome 430A Array 1422303_PM_a_at - -
Affymetrix HT Mouse Genome 430A Array 1422303_a_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 3170746 - -
Illumina mouse-6 v1.0 expression beadchip 3170746 - -
Illumina mouse-6 v1.0 expression beadchip 430253 - -
Illumina mouseRef-8 v1.0 expression beadchip 0003170746 - -
Illumina mouseRef-8 v1.0 expression beadchip 0000430253 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2465751 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2419490 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2465751 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2419490 - -
mouse-6 v1.1 (Illumina) scl24601.2.1_6-S - -
mouse-6 v1.1 (Illumina) scl0002685.1_109-S - -
Illumina Sentrix mouse-6.v1 bead array 3170746 - -
Illumina Sentrix mouse-6.v1 bead array 430253 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl24601.2.1_6-S - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0002685.1_109-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 430253 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 3170746 - -
MouseRef-8 v2 Expression BeadChip ILMN_2419490 - -
MouseRef-8 v2 Expression BeadChip ILMN_2419494 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2419490 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2419494 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0