This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00437956_m1
See other PDIA4 GE Assays ›
Gene Symbol:
Pdia4
Gene Name:
protein disulfide isomerase associated 4
Gene Aliases:
AI987846, Cai, ERp-72, Erp72
Chromosome Location:
Chr.6: 47796141 - 47813512 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00437956_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pdia4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009787.2 NP_033917.2 2 - 3 467 126
GenBank mRNA AK028292.1 - 2 - 3 317 126
AK146288.1 - 2 - 3 300 126
AK148801.1 - 2 - 3 386 126
AK150566.1 - 2 - 3 348 126
AK161534.1 - 2 - 3 305 126
AK168447.1 - 2 - 3 270 126
AK169387.1 - 2 - 3 312 126
BC066857.1 - 1 - 2 66 126
BC141078.1 - 2 - 3 363 9055690 126
J05186.1 - 2 - 3 350 126
Y00884.1 - 1 - 2 173 126

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600002212.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pdia4

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

protein folding
response to endoplasmic reticulum stress
cell redox homeostasis
chaperone-mediated protein folding
positive regulation of protein folding
protein disulfide isomerase activity
isomerase activity
poly(A) RNA binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600017388.mm.1 NM_009787.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600017388.mm.1 NM_009787.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600017388.mm.1 TC0600002212.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600017390.mm.1 NM_009787.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600017390.mm.1 NM_009787.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600017390.mm.1 TC0600002212.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600017391.mm.1 NM_009787.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600017391.mm.1 NM_009787.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600017391.mm.1 TC0600002212.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600017389.mm.1 NM_009787.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600017389.mm.1 NM_009787.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600017389.mm.1 TC0600002212.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 330017 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline