This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00437957_g1
See other PDIA4 GE Assays ›
Gene Symbol:
Pdia4
Gene Name:
protein disulfide isomerase associated 4
Gene Aliases:
AI987846, Cai, ERp-72, Erp72
Chromosome Location:
Chr.6: 47796141 - 47813512 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00437957_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pdia4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009787.2 NP_033917.2 3 - 4 669 73
GenBank mRNA AK028292.1 - 3 - 4 519 73
AK146288.1 - 3 - 4 502 73
AK148801.1 - 3 - 4 588 73
AK150566.1 - 3 - 4 550 73
AK161534.1 - 3 - 4 507 73
AK168447.1 - 3 - 4 472 73
AK169387.1 - 3 - 4 514 73
BC066857.1 - 2 - 3 268 73
BC141078.1 - 3 - 4 565 9055690 73
J05186.1 - 3 - 4 552 73
Y00884.1 - 2 - 3 375 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600002212.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pdia4

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

protein folding
response to endoplasmic reticulum stress
cell redox homeostasis
chaperone-mediated protein folding
positive regulation of protein folding
protein disulfide isomerase activity
isomerase activity
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600017387.mm.1 NM_009787.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600017387.mm.1 NM_009787.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600017387.mm.1 TC0600002212.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline