This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00437979_g1
See other NR1I3 GE Assays ›
Gene Symbol:
Nr1i3
Gene Name:
nuclear receptor subfamily 1, group I, member 3
Gene Aliases:
AA209988, AI551208, CAR, CAR-beta, Care2, ESTM32, MB67
Chromosome Location:
Chr.1: 171213935 - 171218845 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00437979_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nr1i3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001243062.1 NP_001229991.1 1 - 2 201 66
NM_001243063.1 NP_001229992.1 1 - 2 201 66
NM_009803.5 NP_033933.2 1 - 2 201 66
GenBank mRNA AF009327.1 - 1 - 2 138 66
AF009328.1 - 1 - 2 235 66
AK133515.1 - 1 - 2 202 66
AK149381.1 - 1 - 2 153 66
BC037645.1 - 1 - 2 207 66
BC089541.1 - 1 - 2 150 30315989 66

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF009327.1, AF009328.1, AK133515.1, BC037645.1, BC089541.1, NM_001243062.1, NM_001243063.1, NM_009803.5 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nr1i3

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
steroid hormone mediated signaling pathway
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
steroid hormone receptor activity
thyroid hormone receptor activity
protein binding
zinc ion binding
sequence-specific DNA binding
metal ion binding

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 916213 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0