This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00437986_m1
See other NR1I3 GE Assays ›
Gene Symbol:
Nr1i3
Gene Name:
nuclear receptor subfamily 1, group I, member 3
Gene Aliases:
AA209988, AI551208, CAR, CAR-beta, Care2, ESTM32, MB67
Chromosome Location:
Chr.1: 171213935 - 171218845 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00437986_m1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nr1i3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001243063.1 NP_001229992.1 8 - 9 1150 79
NM_009803.5 NP_033933.2 8 - 9 1153 79
GenBank mRNA AF009327.1 - 8 - 9 1090 79
AK133515.1 - 8 - 9 1154 79
AK149430.1 - 864 79
BC037645.1 - 6 - 7 1223 79
BC089541.1 - 8 - 9 1099 30315989 79

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100001579.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Nuclear Receptor 3' Most Mammalian Gene Collection Mus musculus Mouse Estrogen Panel Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nr1i3

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
steroid hormone mediated signaling pathway
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
steroid hormone receptor activity
thyroid hormone receptor activity
protein binding
zinc ion binding
sequence-specific DNA binding
metal ion binding

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20826591 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20826590 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20826588 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20826589 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2035824 BC058978.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2035824 AK140721.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2035824 AK028388.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2035824 NM_001037170.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2035824 BC037645.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P358256 AK140721.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P358256 BC037645.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P358256 BC089541.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P358256 AK149381.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P358256 AK149430.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P358256 AK140721.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P358256 BC037645.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P358256 BC089541.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P358256 AK149381.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P358256 AK149430.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 256861 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 673458 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 532210 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P179523 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P358256 AK140721.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P358256 BC037645.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P358256 BC089541.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P358256 AK149381.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P358256 AK149430.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P358256 AK140721.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P358256 BC037645.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P358256 BC089541.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P358256 AK149381.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P358256 AK149430.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2035824 BC058978.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2035824 AK140721.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2035824 AK028388.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2035824 NM_001037170.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2035824 BC037645.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P358256 AK140721.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P358256 BC037645.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P358256 BC089541.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P358256 AK149381.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P358256 AK149430.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P358256 AK140721.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P358256 BC037645.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P358256 BC089541.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P358256 AK149381.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P358256 AK149430.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20826664 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20826666 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20826635 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20826660 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20826654 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20826659 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20826645 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20826641 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20826662 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20826647 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20826637 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20826651 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20826642 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20826643 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20826636 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20826657 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20826639 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20826656 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20826663 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P179523 - -
Whole Mouse Genome G4122A A_51_P358256 AK140721.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P358256 BC037645.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P358256 BC089541.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P358256 AK149381.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P358256 AK149430.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P358256 AK140721.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P358256 BC037645.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P358256 BC089541.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P358256 AK149381.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P358256 AK149430.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121510 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P179523 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00641543 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00921937 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00641553 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00725254 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00641555 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00641546 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00641547 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20826588 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20826591 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20826590 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20826589 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826747 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826720 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826713 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826741 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826755 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826748 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826721 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826668 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826715 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826652 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826657 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826673 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826647 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826626 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826625 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826670 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826712 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826662 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826694 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826660 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826732 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826744 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826639 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826753 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826637 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826675 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826729 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826696 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826707 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826710 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826681 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826666 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826724 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826759 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826677 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826693 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826663 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826614 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826749 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826635 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826659 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826757 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826623 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826643 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826654 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826683 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826620 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826739 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826717 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826646 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826703 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826679 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826612 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826751 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826726 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826728 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826737 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826691 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826705 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826610 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826746 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826656 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826702 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826709 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20826683 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00632306 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00360180 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20826681 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20826636 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00023051 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20826670 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20826647 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20826682 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20826630 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20826657 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20826621 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00868415 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20826637 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00641543 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00946063 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20826675 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20826677 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00868364 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20826646 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20826643 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20826633 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20826654 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20826663 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20826619 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20826666 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20826618 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00303455 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20826661 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20826656 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20826635 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20826660 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00868406 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00600894 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00725254 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20826639 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00641556 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20826672 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20826679 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10351544 - -
Affymetrix Mouse Gene 1.0 ST Array 10351536 - -
Affymetrix Mouse Gene 1.0 ST Array 10351535 - -
Affymetrix Mouse Gene 1.0 ST Array 10351540 - -
Affymetrix Mouse Gene 1.0 ST Array 10351537 - -
Affymetrix Mouse Gene 1.0 ST Array 10351538 - -
Affymetrix Mouse Gene 1.0 ST Array 10351545 - -
Affymetrix Mouse Gene 1.0 ST Array 10351542 - -
Affymetrix Mouse Gene 1.0 ST Array 10351534 - -
Affymetrix Mouse Gene 1.0 ST Array 10351543 - -
Affymetrix Mouse Gene 1.0 ST Array 10351539 - -
Affymetrix HT MG-430 PM Array Plate 1425392_PM_a_at - -
Affymetrix Mouse Gene 1.1 ST Array 10351533 - -
Mouse Genome 430 2.0 Array 1455614_at - -
Mouse Genome 430 2.0 Array 1429382_at - -
Mouse Genome 430 2.0 Array 1455614_PM_at - -
Mouse Genome 430 2.0 Array 1429382_PM_at - -
Mouse Genome 430 2.0 Array 1425392_PM_a_at - -
Mouse Genome 430 2.0 Array 1425392_a_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 12355_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 12355_at - -
Affymetrix Mouse Clariom S Array PSR0100012940.mm.1 NM_001243063.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100012940.mm.1 NM_001243063.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100012940.mm.1 NM_009803.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100012940.mm.1 NM_009803.5 Refseq
Affymetrix Mouse Clariom S Array PSR0100012940.mm.1 TC0100001579.mm.1 MTA TC
Mouse Expression Set 430 1425392_PM_a_at - -
Mouse Expression Set 430 1425392_a_at - -
Murine Genome U74v2 Set 104506_at - -
Murine Genome U74v2 Set 102171_r_at - -
Murine Genome U74v2 Set 104507_PM_g_at - -
Murine Genome U74v2 Set 104507_g_at - -
Murine Genome U74v2 Set 104506_PM_at - -
Murine Genome U74v2 Set 102171_PM_r_at - -
Mouse Genome 430A 2.0 Array 1425392_a_at - -
Mouse Genome 430A 2.0 Array 1425392_PM_a_at - -
Affymetrix HT Mouse Genome 430A Array 1425392_PM_a_at - -
Affymetrix HT Mouse Genome 430A Array 1425392_a_at - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 50026 - -
Illumina mouse-6 v1.0 expression beadchip 1240673 - -
Illumina mouseRef-8 v1.0 expression beadchip 0001240673 - -
Illumina mouseRef-8 v1.0 expression beadchip 0000050026 - -
mouse-6 v1.1 (Illumina) scl000059.1_14-S - -
Illumina Sentrix mouse-6.v1 bead array 50026 - -
Illumina Sentrix mouse-6.v1 bead array 1240673 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl000059.1_14-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 1240673 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 50026 - -
MouseRef-8 v2 Expression BeadChip ILMN_2926661 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2588846 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1212992 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2926661 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0