This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00438807_m1
See other EYA3 GE Assays ›
Gene Symbol:
Eya3
Gene Name:
eyes absent 3 homolog (Drosophila)
Gene Aliases:
AI844637
Chromosome Location:
Chr.4: 132639016 - 132724765 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00438807_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eya3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010166.3 NP_034296.2 5 - 6 367 63
NM_210071.2 NP_997592.1 4 - 5 328 63
NM_211357.2 NP_997596.1 3 - 4 284 63
GenBank mRNA AK144653.1 - 4 - 5 319 63
AK164591.1 - 4 - 5 328 63
BC052860.1 - 4 - 5 307 63
BC063259.1 - 4 - 5 256 6406090 63
BY102452.1 - 4 - 5 320 63
BY730487.1 - 4 - 5 328 63
U81604.1 - 5 - 6 335 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400001594.mm.1

Additional Information:

This assay is in the 5'UTR region of the NM_211357.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eya3

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
double-strand break repair
transcription, DNA-templated
regulation of transcription, DNA-templated
protein dephosphorylation
cellular response to DNA damage stimulus
multicellular organism development
response to ionizing radiation
chromatin modification
histone dephosphorylation
peptidyl-tyrosine dephosphorylation
positive regulation of DNA repair
chromatin binding
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
protein binding
protein tyrosine/serine/threonine phosphatase activity
hydrolase activity
metal ion binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400012513.mm.1 NM_010166.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400012513.mm.1 NM_210071.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400012513.mm.1 TC0400001594.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400012511.mm.1 NM_010166.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400012511.mm.1 NM_210071.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400012511.mm.1 NM_211357.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400012511.mm.1 TC0400001594.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400012508.mm.1 NM_010166.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400012508.mm.1 NM_210071.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400012508.mm.1 TC0400001594.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400012506.mm.1 NM_010166.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400012506.mm.1 TC0400001594.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 364025 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jmd5k:80/100.66.78.41:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0