This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00438823_m1
See other EYA4 GE Assays ›
Gene Symbol:
Eya4
Gene Name:
eyes absent 4 homolog (Drosophila)
Gene Aliases:
B130023L16Rik
Chromosome Location:
Chr.10: 23102962 - 23350856 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00438823_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eya4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010167.4 NP_034297.2 3 - 4 288 64
GenBank mRNA AK041625.1 - 3 - 4 289 64
BC120898.1 - 3 - 4 272 40061597 64
BC120899.1 - 3 - 4 272 40061599 64
Y17115.1 - 4 - 5 399 64

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eya4

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
transcription, DNA-templated
regulation of transcription, DNA-templated
cellular response to DNA damage stimulus
multicellular organism development
sensory perception of sound
chromatin modification
middle ear morphogenesis
molecular_function
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
hydrolase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0