This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00439045_m1
See other GABRA1 GE Assays ›
Gene Symbol:
Gabra1
Gene Name:
gamma-aminobutyric acid (GABA) A receptor, subunit alpha 1
Gene Aliases:
Gabra-1
Chromosome Location:
Chr.11: 42130939 - 42182951 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00439045_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gabra1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010250.5 NP_034380.1 8 - 9 1633 77
GenBank mRNA AK034095.1 - 8 - 9 1634 77
AK039496.1 - 8 - 9 917 77
AK141596.1 - 8 - 9 1493 77
BC132331.1 - 8 - 9 1309 40130608 77
BC137733.1 - 8 - 9 1312 8860752 77
M63436.1 - 8 - 9 1306 77
M86566.1 - 8 - 9 1001 77
X61430.1 - 7 - 8 889 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100002557.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gabra1

Panther Classification:

Molecular Function -

ligand-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
chloride transport
gamma-aminobutyric acid signaling pathway
synaptic transmission, GABAergic
cellular response to histamine
GABA-A receptor activity
ion channel activity
extracellular ligand-gated ion channel activity
chloride channel activity
protein binding
drug binding
GABA receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline