This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00439105_m1
See other GJA1 GE Assays ›
Gene Symbol:
Gja1
Gene Name:
gap junction protein, alpha 1
Gene Aliases:
AU042049, AW546267, Cnx43, Cx43, Cx43alpha1, Gja-1, Npm1, connexin43
Chromosome Location:
Chr.10: 56377201 - 56390423 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00439105_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gja1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010288.3 NP_034418.1 1 - 2 220 65
GenBank mRNA AK011459.1 - 1 - 2 201 65
AK029291.1 - 1 - 2 77 65
AK036979.1 - 1 - 2 191 65
AK145692.1 - 1 - 2 190 65
AK165986.1 - 1 - 2 77 65
AK171955.1 - 1 - 2 189 65
AY427555.1 - 1 - 2 247 65
BC006894.1 - 1 - 2 173 3599347 65
BC055375.1 - 1 - 2 143 30063597 65
BY061175.1 - 1 - 2 223 65
M63801.1 - 1 - 2 208 65
X61576.1 - 1 - 2 74 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000000493.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK011459.1,AK029291.1,AK036979.1,AK145692.1,AK165986.1,AY427555.1,BC006894.1,BC055375.1,M63801.1,NM_010288.3,X61576.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gja1

Panther Classification:

Molecular Function -

gap junction

Gene Ontology Categories:

Function(s) Process(es)

osteoblast differentiation
in utero embryonic development
neuron migration
negative regulation of endothelial cell proliferation
heart looping
regulation of heart rate
epithelial cell maturation
lens development in camera-type eye
chronic inflammatory response
positive regulation of glomerular filtration
atrial ventricular junction remodeling
epicardial cell to mesenchymal cell transition
apoptotic process
cell communication
signal transduction
positive regulation of cytosolic calcium ion concentration
cell-cell signaling
heart development
adult heart development
regulation of heart contraction
negative regulation of cell proliferation
response to pH
vascular transport
positive regulation of gene expression
negative regulation of gene expression
cell communication by chemical coupling
cell communication by electrical coupling
positive regulation of cell communication by chemical coupling
ATP transport
regulation of bone mineralization
positive regulation of insulin secretion
ion transmembrane transport
embryonic heart tube development
embryonic digit morphogenesis
positive regulation of I-kappaB kinase/NF-kappaB signaling
skeletal muscle tissue regeneration
cell-cell junction organization
positive regulation of osteoblast differentiation
positive regulation of protein catabolic process
positive regulation of striated muscle tissue development
positive regulation of vasoconstriction
positive regulation of vasodilation
regulation of bone remodeling
blood vessel morphogenesis
neuron projection morphogenesis
protein oligomerization
regulation of calcium ion transport
transmembrane transport
negative regulation of cardiac muscle cell proliferation
milk ejection
regulation of ventricular cardiac muscle cell membrane repolarization
regulation of blood vessel remodeling
regulation of atrial cardiac muscle cell membrane depolarization
regulation of ventricular cardiac muscle cell membrane depolarization
negative regulation of wound healing
cardiac conduction
establishment of epithelial cell polarity
negative regulation of DNA biosynthetic process
regulation of bicellular tight junction assembly
positive regulation of behavioral fear response
signal transducer activity
receptor binding
gap junction channel activity
protein binding
ion transmembrane transporter activity
SH3 domain binding
protein domain specific binding
transmembrane transporter activity
PDZ domain binding
beta-tubulin binding
connexin binding
scaffold protein binding
gap junction channel activity involved in cell communication by electrical coupling

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 731884 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline