This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00439155_m1
See other GSR GE Assays ›
Gene Symbol:
Gsr
Gene Name:
glutathione reductase
Gene Aliases:
AI325518, D8Ertd238e, Gr-1, Gr1
Chromosome Location:
Chr.8: 33653238 - 33698162 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00439155_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gsr
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010344.4 NP_034474.4 9 - 10 1276 74
GenBank mRNA AK040136.1 - 9 - 10 985 74
AK084328.1 - 9 - 10 906 74
AK151063.1 - 9 - 10 922 74
AK151247.1 - 9 - 10 922 74
AK151674.1 - 9 - 10 834 74
AK152873.1 - 9 - 10 925 74
AK159678.1 - 9 - 10 948 74
AK167393.1 - 9 - 10 907 74
AK196810.1 - 2 - 3 186 74
BC006966.1 - 6 - 7 601 74
BC056357.1 - 9 - 10 1276 6813295 74
BC057325.1 - 9 - 10 1276 6813295 74
BQ178602.1 - 6 - 7 586 74
X76341.2 - 9 - 10 974 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800000394.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gsr

Panther Classification:

Molecular Function -

reductase

Gene Ontology Categories:

Function(s) Process(es)

glutathione metabolic process
spermatogenesis
cell redox homeostasis
oxidation-reduction process
glutathione-disulfide reductase activity
oxidoreductase activity
oxidoreductase activity, acting on a sulfur group of donors, NAD(P) as acceptor
protein homodimerization activity
glutathione binding
flavin adenine dinucleotide binding
NADP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline