This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00440568_m1
See other OPRM1 GE Assays ›
Gene Symbol:
Oprm1
Gene Name:
opioid receptor, mu 1
Gene Aliases:
M-OR-1, MOP-R, MOR-1, MOR-1O, Oprm, mor, muOR
Chromosome Location:
Chr.10: 6758593 - 7038209 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00440568_m1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information
  • Citations (1)

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Oprm1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AF074974.1 - 4 - 5 1343 73
AF167568.1 - 5 - 6 1529 73
AY358080.1 - 3 - 4 942 73
DQ363374.1 - 2 - 3 224 73
DQ868787.1 - 5 - 6 1744 73
DQ868788.2 - 5 - 6 73
EF105311.1 - 4 - 5 1343 73
EF105312.1 - 4 - 5 1343 73
EF105313.1 - 5 - 6 1645 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mus musculus Inventoried Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor transmembrane signal receptor

Back To Top
 

Citations (1)

Tolerance to the antinociceptive and antiexudative effects of morphine in a murine model of peripheral inflammation.
Authors
Fernandez-Duenas V, Pol O, Garcia-Nogales P, Hernandez L, Planas E, Puig MM
Journal
J Pharmacol Exp Ther - Page No 360 (Sun Jul 01 00:00:00 EDT 2007)
Volume (Year)
322 (2007)
Product Usage
-

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0