This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00440597_g1
See other P2RX6 GE Assays ›
Gene Symbol:
P2rx6
Gene Name:
purinergic receptor P2X, ligand-gated ion channel, 6
Gene Aliases:
P2rxl1, P2x6, P2xm
Chromosome Location:
Chr.16: 17561855 - 17572012 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00440597_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: P2rx6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001159561.1 NP_001153033.1 8 - 9 1041 77
NM_011028.2 NP_035158.2 9 - 10 1122 77
GenBank mRNA AB010883.1 - 9 - 10 1007 77
AK054195.1 - 965 77
AK081308.1 - 9 - 10 999 77
AK138541.1 - 9 - 10 974 77
BC117713.2 - 8 - 9 903 40088045 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600000220.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene P2rx6

Panther Classification:

Molecular Function -

ligand-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
cation transport
response to ATP
ion transmembrane transport
purinergic nucleotide receptor signaling pathway
protein homooligomerization
protein heterooligomerization
cation transmembrane transport
purinergic nucleotide receptor activity
extracellular ATP-gated cation channel activity
ion channel activity
ATP binding
identical protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600001891.mm.1 NM_001159561.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600001891.mm.1 NM_001159561.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600001891.mm.1 NM_011028.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600001891.mm.1 NM_011028.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600001891.mm.1 TC1600000220.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0