This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00440974_m1
See other EIF2AK2 GE Assays ›
Gene Symbol:
Eif2ak2
Gene Name:
eukaryotic translation initiation factor 2-alpha kinase 2
Gene Aliases:
2310047A08Rik, 4732414G15Rik, AI467567, AI747578, Pkr, Prkr, Tik
Chromosome Location:
Chr.17: 78852550 - 78882572 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00440974_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eif2ak2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011163.4 NP_035293.1 9 - 10 894 98
GenBank mRNA AK028602.1 - 9 - 10 894 98
BC016422.1 - 9 - 10 857 3964935 98
M65029.1 - 9 - 10 846 98
M93567.1 - 9 - 10 836 98

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700002558.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eif2ak2

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

activation of MAPKK activity
positive regulation of cytokine production
immune system process
transcription, DNA-templated
regulation of transcription, DNA-templated
translation
protein phosphorylation
response to virus
response to toxic substance
phosphorylation
negative regulation of translation
endoplasmic reticulum unfolded protein response
response to lipopolysaccharide
positive regulation of chemokine production
positive regulation of stress-activated MAPK cascade
response to vitamin E
negative regulation of osteoblast proliferation
response to interferon-alpha
positive regulation of apoptotic process
negative regulation of apoptotic process
negative regulation of viral genome replication
innate immune response
protein autophosphorylation
positive regulation of NF-kappaB transcription factor activity
defense response to virus
regulation of NLRP3 inflammasome complex assembly
positive regulation of NIK/NF-kappaB signaling
regulation of hematopoietic progenitor cell differentiation
regulation of hematopoietic stem cell proliferation
regulation of hematopoietic stem cell differentiation
nucleotide binding
RNA binding
double-stranded RNA binding
protein kinase activity
protein serine/threonine kinase activity
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0