This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00441675_g1
See other TAP2 GE Assays ›
Gene Symbol:
Tap2
Gene Name:
transporter 2, ATP-binding cassette, sub-family B (MDR/TAP)
Gene Aliases:
ABC18, AI462429, APT2, Abcb3, Ham-2, Ham2, MTP2, PSF2, RING11, Tap-2, Y1, jas
Chromosome Location:
Chr.17: 34204085 - 34216321 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00441675_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tap2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011530.3 NP_035660.3 10 - 11 1947 104
GenBank mRNA AK020022.1 - 9 - 10 2023 104
AK079667.1 - 10 - 11 1861 104
AK145234.1 - 10 - 11 1906 104
AK151376.1 - 10 - 11 1857 104
AK151483.1 - 10 - 11 1857 104
AK151657.1 - 10 - 11 1880 104
AK153287.1 - 10 - 11 1857 104
AK159200.1 - 10 - 11 1913 104
AK161091.1 - 10 - 11 1863 104
BC005578.1 - 10 - 11 1847 3968225 104
BC051257.1 - 10 - 11 1860 6530488 104
BC057668.1 - 10 - 11 2234 104
CT010387.1 - 9 - 10 1794 104
M90459.1 - 9 - 10 1794 104
U60088.1 - 9 - 10 1797 104
U60089.1 - 9 - 10 1797 104
U60090.1 - 9 - 10 1797 104
U60091.1 - 9 - 10 1797 104
U60092.1 - 9 - 10 1794 104

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tap2

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of T cell mediated cytotoxicity
response to molecule of bacterial origin
immune system process
antigen processing and presentation of exogenous protein antigen via MHC class Ib, TAP-dependent
antigen processing and presentation of endogenous peptide antigen via MHC class I via ER pathway, TAP-dependent
antigen processing and presentation of endogenous peptide antigen via MHC class Ib via ER pathway, TAP-dependent
positive regulation of antigen processing and presentation of peptide antigen via MHC class I
protein complex assembly
transport
protein transport
peptide transport
intracellular transport of viral protein in host cell
antigen processing and presentation of endogenous peptide antigen via MHC class I
protection from natural killer cell mediated cytotoxicity
cytosol to ER transport
peptide antigen transport
transmembrane transport
nucleotide binding
protein binding
ATP binding
peptide transporter activity
peptide antigen-transporting ATPase activity
peptide-transporting ATPase activity
ATPase activity
MHC class Ib protein binding
MHC protein binding
MHC class I protein binding
peptide antigen binding
ATPase activity, coupled to transmembrane movement of substances
protein homodimerization activity
ADP binding
TAP1 binding
TAP2 binding
tapasin binding
protein heterodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0