This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00442537_m1
See other SLC7A11 GE Assays ›
Gene Symbol:
Slc7a11
Gene Name:
solute carrier family 7 (cationic amino acid transporter, y+ system), member 11
Gene Aliases:
9930009M05Rik, AI451155, sut, xCT
Chromosome Location:
Chr.3: 50364936 - 50499087 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00442537_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc7a11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011990.2 NP_036120.1 9 - 10 1472 58
GenBank mRNA AB022345.1 - 9 - 10 1439 58
AK076322.1 - 9 - 10 1447 58
AK089202.1 - 9 - 10 1445 58
AK162233.1 - 9 - 10 1247 58
AK163168.1 - 9 - 10 1446 58
AK165599.1 - 9 - 10 1448 58
AK171192.1 - 9 - 10 1470 58
AK171252.1 - 9 - 10 1446 58
AY766236.1 - 9 - 10 1439 58
AY766237.1 - 1439 58
BC141402.1 - 9 - 10 1369 9056015 58
BC141408.1 - 9 - 10 1369 9056021 58

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300001937.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc7a11

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

amino acid transmembrane transport
transport
amino acid transport
response to toxic substance
neutral amino acid transport
platelet aggregation
modified amino acid transport
L-glutamate transmembrane transport
amino acid transmembrane transporter activity
cystine:glutamate antiporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0