This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00443504_g1
See other TNFRSF19 GE Assays ›
Gene Symbol:
Tnfrsf19
Gene Name:
tumor necrosis factor receptor superfamily, member 19
Gene Aliases:
AL023044, AW123854, TAJ, TAJ-ALPHA, TRADE, Troy
Chromosome Location:
Chr.14: 60963834 - 61046855 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00443504_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tnfrsf19
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001164155.1 NP_001157627.1 2 - 3 233 64
NM_013869.5 NP_038897.4 2 - 3 836 64
GenBank mRNA AB040432.1 - 2 - 3 197 64
AB040433.1 - 2 - 3 137 64
AF167552.1 - 2 - 3 142 64
AF167553.1 - 2 - 3 142 64
AF167554.1 - 2 - 3 165 64
AK011232.1 - 2 - 3 481 64
AK028146.1 - 2 - 3 143 64
AK083283.1 - 2 - 3 451 64
BB858206.1 - 2 - 3 235 64
BC030062.1 - 2 - 3 471 5256883 64
CB244999.1 - 2 - 3 232 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400002180.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tnfrsf19

Gene Ontology Categories:

Function(s)

hair follicle development
positive regulation of I-kappaB kinase/NF-kappaB signaling

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400016894.mm.1 NM_013869.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400016894.mm.1 NM_013869.5 Refseq
Affymetrix Mouse Clariom S Array PSR1400016894.mm.1 TC1400002180.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400016895.mm.1 NM_013869.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400016895.mm.1 NM_013869.5 Refseq
Affymetrix Mouse Clariom S Array PSR1400016895.mm.1 TC1400002180.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400016891.mm.1 NM_001164155.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400016891.mm.1 NM_001164155.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400016891.mm.1 NM_013869.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400016891.mm.1 NM_013869.5 Refseq
Affymetrix Mouse Clariom S Array PSR1400016891.mm.1 TC1400002180.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0