This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00443508_m1
See other TNFRSF19 GE Assays ›
Gene Symbol:
Tnfrsf19
Gene Name:
tumor necrosis factor receptor superfamily, member 19
Gene Aliases:
AL023044, AW123854, TAJ, TAJ-ALPHA, TRADE, Troy
Chromosome Location:
Chr.14: 60963834 - 61046855 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00443508_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tnfrsf19
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001164155.1 NP_001157627.1 6 - 7 777 77
NM_013869.5 NP_038897.4 6 - 7 1380 77
GenBank mRNA AB040432.1 - 6 - 7 741 77
AF167552.1 - 6 - 7 686 77
AF173166.1 - 5 - 6 77
AK028146.1 - 6 - 7 687 77
AK050262.1 - 2 - 3 346 77
AK083283.1 - 6 - 7 995 77

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tnfrsf19

Gene Ontology Categories:

Function(s)

hair follicle development
positive regulation of I-kappaB kinase/NF-kappaB signaling

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0