This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00443665_m1
See other TXNRD1 GE Assays ›
Gene Symbol:
Txnrd1
Gene Name:
thioredoxin reductase 1
Gene Aliases:
TR, TR1, TrxR1
Chromosome Location:
Chr.10: 82833951 - 82897724 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00443665_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Txnrd1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001042513.1 NP_001035978.1 4 - 5 401 65
NM_001042514.1 NP_001035979.1 3 - 4 489 65
NM_001042523.1 NP_001035988.1 4 - 5 618 65
NM_015762.2 NP_056577.2 3 - 4 294 65
GenBank mRNA AB027565.1 - 3 - 4 321 65
AF333036.1 - 4 - 5 618 65
AK011902.1 - 4 - 5 402 65
AK146125.1 - 3 - 4 295 65
AK149625.1 - 4 - 5 402 65
AK151927.1 - 4 - 5 402 65
AK168356.1 - 4 - 5 402 65
BC037643.1 - 3 - 4 574 5059831 65
CJ136679.1 - 4 - 5 402 65
CX562215.1 - 3 - 4 489 65

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Txnrd1

Panther Classification:

Molecular Function -

reductase

Gene Ontology Categories:

Function(s) Process(es)

mesoderm formation
response to oxidative stress
gastrulation
cell proliferation
positive regulation of cell death
selenocysteine metabolic process
benzene-containing compound metabolic process
hydrogen peroxide catabolic process
cell redox homeostasis
protein tetramerization
oxidation-reduction process
NADPH oxidation
thioredoxin-disulfide reductase activity
NAD(P)H oxidase activity
oxidoreductase activity
oxidoreductase activity, acting on a sulfur group of donors, NAD(P) as acceptor
selenate reductase activity
protein homodimerization activity
mercury ion binding
flavin adenine dinucleotide binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0