This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00444145_m1
See other CCS GE Assays ›
Gene Symbol:
Ccs
Gene Name:
copper chaperone for superoxide dismutase
Gene Aliases:
Ccsd
Chromosome Location:
Chr.19: 4825366 - 4839327 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00444145_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ccs
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_016892.3 NP_058588.1 2 - 3 199 61
GenBank mRNA AF121906.1 - 2 - 3 112 61
AF173379.1 - 2 - 3 172 61
AK050072.1 - 2 - 3 148 61
BC026938.1 - 2 - 3 166 4024140 61
BY073478.1 - 2 - 3 199 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900000937.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ccs

Panther Classification:

Molecular Function -

oxidoreductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

superoxide metabolic process
removal of superoxide radicals
metal ion transport
positive regulation of oxidoreductase activity
oxidation-reduction process
superoxide dismutase activity
copper ion binding
zinc ion binding
enzyme binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900008252.mm.1 NM_016892.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900008252.mm.1 NM_016892.3 Refseq
Affymetrix Mouse Clariom S Array PSR1900008252.mm.1 TC1900000937.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0