This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00444444_g1
See other NME6 GE Assays ›
Gene Symbol:
Nme6
Gene Name:
NME/NM23 nucleoside diphosphate kinase 6
Gene Aliases:
nm23-M6
Chromosome Location:
Chr.9: 109832711 - 109842974 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00444444_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nme6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_018757.1 NP_061227.1 2 - 3 215 96
GenBank mRNA AF051942.1 - 2 - 3 215 96
BC002007.1 - 2 - 3 215 3593804 96

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900001441.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nme6

Panther Classification:

Molecular Function -

kinase

Gene Ontology Categories:

Function(s) Process(es)

nucleoside diphosphate phosphorylation
GTP biosynthetic process
UTP biosynthetic process
CTP biosynthetic process
nucleotide metabolic process
phosphorylation
negative regulation of cell growth
negative regulation of mitotic nuclear division
nucleotide binding
nucleoside diphosphate kinase activity
ATP binding
kinase activity
transferase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900011971.mm.1 NM_018757.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900011971.mm.1 NM_018757.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900011971.mm.1 TC0900001441.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline