This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00444456_g1
See other APBA3 GE Assays ›
Gene Symbol:
Apba3
Gene Name:
amyloid beta (A4) precursor protein-binding, family A, member 3
Gene Aliases:
AW208791, Mint-3, X11gamma, lin-10, mint3
Chromosome Location:
Chr.10: 81266965 - 81273247 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00444456_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Apba3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_018758.2 NP_061228.1 9 - 10 1775 95
GenBank mRNA AF070975.1 - 9 - 10 1729 95
AK045965.1 - 9 - 10 1776 95
AK082046.1 - 1695 95
AK169462.1 - 9 - 10 1759 95
AK170182.1 - 9 - 10 1723 95
BC005423.1 - 9 - 10 1707 3487448 95
BC009666.1 - 9 - 10 1719 3490710 95
BQ031518.1 - 2 - 3 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000000891.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Apba3

Panther Classification:

Molecular Function -

membrane trafficking regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
transport
chemical synaptic transmission
regulation of gene expression
protein transport
negative regulation of catalytic activity
beta-amyloid binding
enzyme inhibitor activity
enzyme binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000006985.mm.1 NM_018758.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000006985.mm.1 NM_018758.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000006985.mm.1 TC1000000891.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-tks2p:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0